Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011065.1 | NP_001011065.1 | 2 - 3 | 533 | 80 | |
GenBank mRNA | BC084445.1 | - | 2 - 3 | 533 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |