Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011068.1 | NP_001011068.1 | 7 - 8 | 750 | 82 | |
GenBank mRNA | BC084448.1 | - | 7 - 8 | 750 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |