Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mob3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011080.1 | NP_001011080.1 | 2 - 3 | 618 | 111 | |
XM_012960681.2 | XP_012816135.1 | 3 - 4 | 1140 | 111 | ||
GenBank mRNA | BC084470.1 | - | 2 - 3 | 618 | 111 | |
CT025201.2 | - | 862 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |