Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011094.1 | NP_001011094.1 | 8 - 9 | 1074 | 55 | |
GenBank mRNA | BC084487.1 | - | 8 - 9 | 1074 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |