Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011096.1 | NP_001011096.1 | 3 - 4 | 372 | 68 | |
GenBank mRNA | BC084490.1 | - | 3 - 4 | 372 | 68 | |
CR760770.2 | - | 3 - 4 | 314 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |