Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011107.1 | NP_001011107.1 | 2 - 3 | 511 | 63 | |
GenBank mRNA | BC084504.1 | - | 2 - 3 | 511 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |