Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011120.1 | NP_001011120.1 | 4 - 5 | 434 | 90 | |
XM_012962931.2 | XP_012818385.1 | 4 - 5 | 485 | 90 | ||
XM_012962932.2 | XP_012818386.1 | 4 - 5 | 443 | 90 | ||
XM_012962933.1 | XP_012818387.1 | 4 - 5 | 426 | 90 | ||
XM_012962934.2 | XP_012818388.1 | 4 - 5 | 417 | 90 | ||
XM_012962935.2 | XP_012818389.1 | 4 - 5 | 475 | 90 | ||
XM_012962936.1 | XP_012818390.1 | 4 - 5 | 623 | 90 | ||
GenBank mRNA | BC084519.1 | - | 4 - 5 | 434 | 90 | |
CR760319.2 | - | 3 - 4 | 377 | 90 |
Gene Symbol: mob3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CT025201.2 | - | 4212 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |