Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nutf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011126.1 | NP_001011126.1 | 4 - 5 | 326 | 113 | |
GenBank mRNA | BC084526.1 | - | 4 - 5 | 326 | 113 | |
CR760342.2 | - | 3 - 4 | 315 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |