Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011139.1 | NP_001011139.1 | 9 - 10 | 1087 | 95 | |
GenBank mRNA | BC084468.1 | - | 9 - 10 | 1087 | 95 | |
CR855769.2 | - | 9 - 10 | 1086 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |