Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011170.1 | NP_001011170.1 | 2 - 3 | 326 | 103 | |
XM_012960989.2 | XP_012816443.1 | 2 - 3 | 307 | 103 | ||
GenBank mRNA | BC084996.1 | - | 2 - 3 | 326 | 103 | |
CR760886.2 | - | 2 - 3 | 259 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |