Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgfrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011189.1 | NP_001011189.1 | 2 - 3 | 404 | 72 | |
GenBank mRNA | AJ616852.2 | - | 2 - 3 | 404 | 72 | |
BC161343.1 | - | 2 - 3 | 434 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |