Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011194.1 | NP_001011194.1 | 3 - 4 | 471 | 94 | |
GenBank mRNA | BC087608.1 | - | 3 - 4 | 471 | 94 | |
CR760467.2 | - | 3 - 4 | 446 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |