Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ass1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011212.1 | NP_001011212.1 | 7 - 8 | 668 | 114 | |
GenBank mRNA | BC087767.1 | - | 7 - 8 | 668 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |