Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011236.1 | NP_001011236.1 | 3 - 4 | 500 | 79 | |
XM_012971705.1 | XP_012827159.1 | 4 - 5 | 677 | 79 | ||
XM_012971707.2 | XP_012827161.1 | 4 - 5 | 595 | 79 | ||
XM_012971708.1 | XP_012827162.1 | 4 - 5 | 760 | 79 | ||
XM_012971709.2 | XP_012827163.1 | 4 - 5 | 634 | 79 | ||
GenBank mRNA | BC087807.1 | - | 3 - 4 | 500 | 79 | |
CR942701.1 | - | 3 - 4 | 532 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |