Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsd17b13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011240.1 | NP_001011240.1 | 5 - 6 | 723 | 88 | |
GenBank mRNA | BC087812.1 | - | 5 - 6 | 723 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |