Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011242.1 | NP_001011242.1 | 3 - 4 | 283 | 110 | |
XM_012964601.2 | XP_012820055.1 | 3 - 4 | 449 | 110 | ||
XM_018094629.1 | XP_017950118.1 | 3 - 4 | 449 | 110 | ||
XM_018094630.1 | XP_017950119.1 | 3 - 4 | 337 | 110 | ||
XM_018094631.1 | XP_017950120.1 | 3 - 4 | 342 | 110 | ||
GenBank mRNA | BC087814.1 | - | 3 - 4 | 283 | 110 | |
CR761073.2 | - | 3 - 4 | 270 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |