Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btf3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011243.1 | NP_001011243.1 | 2 - 3 | 101 | 76 | |
XM_012960996.2 | XP_012816450.1 | 2 - 3 | 147 | 76 | ||
XM_012960997.2 | XP_012816451.1 | 2 - 3 | 146 | 76 | ||
XM_012960998.2 | XP_012816452.1 | 2 - 3 | 146 | 76 | ||
XM_012960999.2 | XP_012816453.1 | 2 - 3 | 79 | 76 | ||
GenBank mRNA | BC087817.1 | - | 2 - 3 | 101 | 76 | |
CR848347.2 | - | 1 - 2 | 55 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |