Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011255.1 | NP_001011255.1 | 2 - 3 | 219 | 103 | |
XM_012961026.2 | XP_012816480.1 | 3 - 4 | 338 | 103 | ||
XM_012961031.2 | XP_012816485.1 | 3 - 4 | 233 | 103 | ||
GenBank mRNA | BC086500.2 | - | 2 - 3 | 219 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |