Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011255.1 | NP_001011255.1 | 5 - 6 | 464 | 92 | |
XM_012961026.2 | XP_012816480.1 | 6 - 7 | 583 | 92 | ||
XM_012961031.2 | XP_012816485.1 | 6 - 7 | 478 | 92 | ||
GenBank mRNA | BC086500.2 | - | 5 - 6 | 464 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |