Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011258.1 | NP_001011258.1 | 3 - 4 | 396 | 95 | |
XM_012961036.2 | XP_012816490.1 | 3 - 4 | 649 | 95 | ||
GenBank mRNA | BC087569.2 | - | 3 - 4 | 396 | 95 | |
CR761166.2 | - | 3 - 4 | 482 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |