Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cib3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011260.1 | NP_001011260.1 | 2 - 3 | 368 | 87 | |
GenBank mRNA | BC087966.1 | - | 2 - 3 | 368 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |