Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hvcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011262.1 | NP_001011262.1 | 3 - 4 | 377 | 80 | |
XM_012961069.2 | XP_012816523.1 | 3 - 4 | 480 | 80 | ||
XM_012961072.2 | XP_012816526.1 | 3 - 4 | 433 | 80 | ||
GenBank mRNA | BC087968.1 | - | 3 - 4 | 377 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |