Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011268.1 | NP_001011268.1 | 2 - 3 | 346 | 62 | |
GenBank mRNA | BC087977.1 | - | 2 - 3 | 346 | 62 | |
CR760631.2 | - | 2 - 3 | 317 | 62 |
Gene Symbol: LOC101731073 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004920842.1 | XP_004920899.1 | 1 - 2 | 239 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |