Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otud6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011269.1 | NP_001011269.1 | 1 - 2 | 131 | 61 | |
GenBank mRNA | BC087978.1 | - | 1 - 2 | 131 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |