Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myoz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011273.1 | NP_001011273.1 | 6 - 7 | 888 | 89 | |
XM_018092057.1 | XP_017947546.1 | 5 - 6 | 837 | 89 | ||
GenBank mRNA | BC087983.1 | - | 6 - 7 | 888 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |