Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011290.1 | NP_001011290.1 | 5 - 6 | 761 | 88 | |
XM_012964606.2 | XP_012820060.1 | 5 - 6 | 716 | 88 | ||
XM_012964607.2 | XP_012820061.1 | 5 - 6 | 823 | 88 | ||
XM_012964608.2 | XP_012820062.1 | 5 - 6 | 897 | 88 | ||
XM_012964609.2 | XP_012820063.1 | 5 - 6 | 811 | 88 | ||
XM_012964610.2 | XP_012820064.1 | 5 - 6 | 897 | 88 | ||
GenBank mRNA | BC088007.1 | - | 5 - 6 | 761 | 88 | |
CR848357.2 | - | 5 - 6 | 700 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |