Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203568.1 | NP_988899.1 | 1 - 2 | 256 | 79 | |
GenBank mRNA | BC059749.1 | - | 1 - 2 | 256 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |