Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203570.1 | NP_988901.1 | 2 - 3 | 287 | 74 | |
GenBank mRNA | BC061630.1 | - | 2 - 3 | 287 | 74 | |
BC082501.1 | - | 2 - 3 | 357 | 74 | ||
CR761635.2 | - | 2 - 3 | 286 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |