Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akirin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203583.1 | NP_988914.1 | 2 - 3 | 568 | 78 | |
GenBank mRNA | BC061612.1 | - | 2 - 3 | 568 | 78 | |
CR760919.2 | - | 2 - 3 | 380 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |