Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203594.1 | NP_988925.1 | 4 - 5 | 635 | 86 | |
XM_012956636.2 | XP_012812090.1 | 5 - 6 | 1020 | 86 | ||
GenBank mRNA | BC059744.1 | - | 4 - 5 | 635 | 86 | |
CR760793.2 | - | 4 - 5 | 631 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |