Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mak16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203596.1 | NP_988927.1 | 2 - 3 | 129 | 76 | |
GenBank mRNA | BC061599.1 | - | 2 - 3 | 129 | 76 | |
CT030341.1 | - | 2 - 3 | 132 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |