Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203601.1 | NP_988932.1 | 1 - 2 | 614 | 75 | |
GenBank mRNA | BC061421.1 | - | 1 - 2 | 614 | 75 | |
CR762266.2 | - | 1 - 2 | 606 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |