Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011191.2 | NP_001011191.1 | 3 - 4 | 366 | 97 | |
GenBank mRNA | BC087559.2 | - | 3 - 4 | 366 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |