Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011196.1 | NP_001011196.1 | 4 - 5 | 486 | 94 | |
GenBank mRNA | AJ634915.1 | - | 4 - 5 | 486 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |