Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: impact | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011308.1 | NP_001011308.1 | 1 - 2 | 228 | 71 | |
GenBank mRNA | BC088032.1 | - | 1 - 2 | 228 | 71 | |
CR760285.2 | - | 1 - 2 | 235 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |