Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atraid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011316.1 | NP_001011316.1 | 270 | 63 | ||
GenBank mRNA | BC088051.1 | - | 270 | 63 | ||
CU075626.1 | - | 1 - 2 | 266 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |