Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011319.1 | NP_001011319.1 | 7 - 8 | 765 | 91 | |
GenBank mRNA | BC088056.1 | - | 7 - 8 | 765 | 91 | |
CR762255.2 | - | 7 - 8 | 829 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |