Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011322.1 | NP_001011322.1 | 6 - 7 | 680 | 82 | |
GenBank mRNA | BC088068.1 | - | 6 - 7 | 680 | 82 | |
CU075661.1 | - | 6 - 7 | 878 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |