Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011354.1 | NP_001011354.1 | 444 | 88 | ||
GenBank mRNA | BC088518.1 | - | 444 | 88 | ||
CU024992.1 | - | 472 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis |