Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011354.1 | NP_001011354.1 | 737 | 98 | ||
GenBank mRNA | BC088518.1 | - | 737 | 98 | ||
CR760484.2 | - | 586 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis |