Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rspo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011386.1 | NP_001011386.1 | 1 - 2 | 222 | 102 | |
XM_012964623.2 | XP_012820077.1 | 2 - 3 | 1587 | 102 | ||
GenBank mRNA | BC088569.1 | - | 1 - 2 | 222 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |