Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011409.1 | NP_001011409.1 | 5 - 6 | 529 | 110 | |
XM_012956873.2 | XP_012812327.1 | 5 - 6 | 641 | 110 | ||
GenBank mRNA | BC088783.1 | - | 5 - 6 | 529 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |