Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myoz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011415.1 | NP_001011415.1 | 2 - 3 | 128 | 93 | |
XM_012961302.2 | XP_012816756.1 | 3 - 4 | 622 | 93 | ||
XM_012961309.2 | XP_012816763.1 | 3 - 4 | 623 | 93 | ||
XM_012961316.2 | XP_012816770.1 | 2 - 3 | 529 | 93 | ||
GenBank mRNA | BC087984.1 | - | 2 - 3 | 128 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |