Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ela3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011421.1 | NP_001011421.1 | 56 | 96 | ||
GenBank mRNA | BC087729.1 | - | 46 | 96 | ||
BC088036.1 | - | 56 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis |