Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011447.1 | NP_001011447.1 | 1 - 2 | 273 | 74 | |
XM_012968324.2 | XP_012823778.1 | 1 - 2 | 520 | 74 | ||
GenBank mRNA | BC088535.1 | - | 1 - 2 | 273 | 74 | |
CR848483.2 | - | 1 - 2 | 303 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |