Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zcchc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011465.1 | NP_001011465.1 | 2 - 3 | 488 | 107 | |
GenBank mRNA | BC088591.1 | - | 2 - 3 | 488 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |