Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011480.1 | NP_001011480.1 | 3 - 4 | 216 | 100 | |
GenBank mRNA | BC088779.1 | - | 3 - 4 | 216 | 100 | |
CR761832.2 | - | 3 - 4 | 196 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |