Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult1c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011489.1 | NP_001011489.1 | 2 - 3 | 269 | 59 | |
XM_012954435.1 | XP_012809889.1 | 8 - 9 | 1050 | 59 | ||
XM_018089761.1 | XP_017945250.1 | 8 - 9 | 1050 | 59 | ||
GenBank mRNA | BC088870.1 | - | 2 - 3 | 269 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |