Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011495.1 | NP_001011495.1 | 5 - 6 | 457 | 96 | |
XM_012961488.1 | XP_012816942.1 | 5 - 6 | 454 | 96 | ||
XM_018092899.1 | XP_017948388.1 | 5 - 6 | 1290 | 96 | ||
XM_018092902.1 | XP_017948391.1 | 4 - 5 | 439 | 96 | ||
GenBank mRNA | BC088885.1 | - | 5 - 6 | 457 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |