Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcaf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015742.2 | NP_001015742.2 | 5 - 6 | 1059 | 85 | |
GenBank mRNA | BC089677.1 | - | 5 - 6 | 1033 | 85 | |
BC159065.1 | - | 5 - 6 | 1047 | 85 | ||
BC159294.1 | - | 5 - 6 | 1038 | 85 | ||
BC170811.1 | - | 5 - 6 | 774 | 85 | ||
BC170813.1 | - | 5 - 6 | 774 | 85 | ||
CR762027.2 | - | 5 - 6 | 959 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |